Learn More
Medchemexpress LLC HY-N4151 5mg Medchemexpress, Chrysophanein CAS:4839-60-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN41515MG
Encompass_Preferred
Medchemexpress, HY-N4151 5mg Chrysophanein CAS:4839-60-5 Chrysophanein is a chrysophanol glycoside from leaves and roots of Aloe hijazensis. Chrysophanein shows a moderate cytotoxic activity against several carcinoma cells lines. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0428