Learn More
Selleck Chemical LLC Chrysophanic Acid 100mg 481-74-3 Chrysophanol
SDP
Supplier:  Selleck Chemical LLC S2406100MG
Encompass_Preferred
Chrysophanic Acid (Chrysophanol) a natural anthraquinone isolated from Dianella longifolia, is a EGFR/mTOR pathway inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-606-2