Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chrysophanol-1-O-β-gentiobioside | 54944-38-6 | 1 MG
SDP

Catalog No. 5000418587
Encompass_Preferred

Chrysophanol-1-O-β-gentiobioside is an anthraquinone glycoside isolated from *Cassia obtusifolia* seeds. It demonstrates selective inhibition of hMAO-A isozyme activity with an IC50 of 96.15 μM.

  • Anthraquinone glycoside
  • Isolated from *Cassia obtusifolia* seeds
  • Selective inhibitor of hMAO-A isozyme activity
  • IC50 of 96.15 μM against MAO-A
  • Molecular weight: 578.52
  • Chemical formula: C27H30O14
  • Appearance: Solid, white to off-white

Catalog No. 50-004-18587 Supplier Medchemexpress LLC Supplier No. HYN75981MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chrysophanol-1-O-β-gentiobioside is an anthraquinone glycoside isolated from *Cassia obtusifolia* seeds. It demonstrates selective inhibition of hMAO-A isozyme activity with an IC50 of 96.15 μM.

  • Anthraquinone glycoside
  • Isolated from *Cassia obtusifolia* seeds
  • Selective inhibitor of hMAO-A isozyme activity
  • IC50 of 96.15 μM against MAO-A
  • Molecular weight: 578.52
  • Chemical formula: C27H30O14
  • Appearance: Solid, white to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.