Learn More
Medchemexpress LLC HY-13595 50mg Medchemexpress, Chrysophanol CAS:481-74-3 Purity:>98%
SDP
Catalog No. 502081414
Encompass_Preferred
Medchemexpress, HY-13595 50mg Chrysophanol CAS:481-74-3 Chrysophanol (Chrysophanic acid) is a natural anthraquinone, which inhibits EGF-induced phosphorylation of EGFR and suppresses activation of AKT and mTOR/p70S6K. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1414 Supplier Medchemexpress LLC Supplier No. HY1359550MG
Medchemexpress, HY-13595 50mg Chrysophanol CAS:481-74-3 Chrysophanol (Chrysophanic acid) is a natural anthraquinone, which inhibits EGF-induced phosphorylation of EGFR and suppresses activation of AKT and mTOR/p70S6K. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.