Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chrysophanol 8-O-glucoside | 14392-02-0 | >98.0% | 432.38 g·mol⁻¹ | C21H20O10 | 10 MG
SDP

Catalog No. 5000289875
Encompass_Preferred

Chrysophanol 8-O-glucoside is a natural anthraquinone glucoside isolated from the roots of Rumex acetosa. It is provided for research and analytical applications and exhibits moderate elastase inhibition activity. Typical preparation is a purified powder with recommended storage conditions for long-term stability.

  • Natural anthraquinone glucoside isolated from Rumex acetosa roots.
  • Shows moderate elastase inhibition activity.
  • High purity suitable for analytical work.
  • Molecular formula C21H20O10; molecular weight 432.38 g·mol⁻¹.
  • Provided as a small research quantity (10 mg) for assay and reference use.
  • Soluble in DMSO; store powder at -20°C for extended stability.

Catalog No. 50-002-89875 Supplier Medchemexpress LLC Supplier No. HYN239510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chrysophanol 8-O-glucoside is a natural anthraquinone glucoside isolated from the roots of Rumex acetosa. It is provided for research and analytical applications and exhibits moderate elastase inhibition activity. Typical preparation is a purified powder with recommended storage conditions for long-term stability.

  • Natural anthraquinone glucoside isolated from Rumex acetosa roots.
  • Shows moderate elastase inhibition activity.
  • High purity suitable for analytical work.
  • Molecular formula C21H20O10; molecular weight 432.38 g·mol⁻¹.
  • Provided as a small research quantity (10 mg) for assay and reference use.
  • Soluble in DMSO; store powder at -20°C for extended stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.