Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chrysophanol, Analytical Standard, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014274 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4274 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4274 Supplier MilliporeSigma Supelco Supplier No. 0154225MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chrysophanol is an anthraquinone compound and occurs naturally in Chinese herb Rheum officinale. It shows antimicrobial activity, and is an active component in many crude drugs having purgative properties.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
CAS 481-74-3
Linear Formula C15H10O4
MDL Number MFCD00001208
Melting Point 192°C to 196°C
Synonym 1,8-Dihydroxy-3-methylanthraquinone; 3-Methylchrysazin; Chrysophanic acid
Quantity 25 mg
Formula Weight 254.24
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.