Learn More
Medchemexpress LLC Chrysosplenol D | 14965-20-9 | MFCD01548059 | 99.9% | 360.31 | 10 MG
SDP
Catalog No. 5000365427
Encompass_Preferred
Chrysosplenol D is a methoxy flavonoid known for inducing ERK1/2-mediated apoptosis in triple-negative human breast cancer cells. It demonstrates anti-inflammatory properties and moderate antitrypanosomal activities.
  • Inhibits cell viability in CaCo2 cells.
  • Selectively inhibits viability of TNBC cell lines.
  • Induces cell cycle aberrations and apoptosis in breast cancer cells.
  • Shows antitrypanosomal activity against T.b. brucei.
  • Inhibits tumor growth on chick chorioallantoic membranes.
  • Protects against LPS-induced systemic inflammatory response syndrome in mice.
  • Inhibits croton oil-induced ear edema in mice.
Catalog No. 50-003-65427 Supplier Medchemexpress LLC Supplier No. 10MGHYN6007
Chrysosplenol D is a methoxy flavonoid known for inducing ERK1/2-mediated apoptosis in triple-negative human breast cancer cells. It demonstrates anti-inflammatory properties and moderate antitrypanosomal activities.
  • Inhibits cell viability in CaCo2 cells.
  • Selectively inhibits viability of TNBC cell lines.
  • Induces cell cycle aberrations and apoptosis in breast cancer cells.
  • Shows antitrypanosomal activity against T.b. brucei.
  • Inhibits tumor growth on chick chorioallantoic membranes.
  • Protects against LPS-induced systemic inflammatory response syndrome in mice.
  • Inhibits croton oil-induced ear edema in mice.