Learn More
Cayman Chemical Chrysosplenetin, Purity: ≥98%, CAS: 603-56-5, 10 mg
SDP
Catalog No. 502991401
Encompass_Preferred
A flavonoid with diverse biological activities; active against P. falciparum in vitro (IC50 = 23 µM); inhibits the cytopathic effect of EV71 in Vero cells (EC50 = 0.68 µM); increases survival in a neonatal mouse model of EV71 infection at 1 and 5 mg/kg; increases proliferation and osteogenic differentiation of isolated BMSCs; prevents estrogen deficiency-induced bone loss in ovariectomized mice
Catalog No. 50-299-1401 Supplier Cayman Chemical Supplier No. 3062510 MG
A flavonoid with diverse biological activities; active against P. falciparum in vitro (IC50 = 23 µM); inhibits the cytopathic effect of EV71 in Vero cells (EC50 = 0.68 µM); increases survival in a neonatal mouse model of EV71 infection at 1 and 5 mg/kg; increases proliferation and osteogenic differentiation of isolated BMSCs; prevents estrogen deficiency-induced bone loss in ovariectomized mice