Learn More
AdipoGen CHS-828 (25 mg)
SDP
Supplier:  AdipoGen AGCR10064M025
Encompass_Preferred
Chemical. CAS: 200484-11-3. Formula: C19H22ClN5O. MW: 371.9. Antitumor compound. Programmed cell death inducer. Cytotoxic. Antiproliferative. p53 activator. IkappaB kinase inhibitor. NF-kappaB inhibitor. Nampt/visfatin inhibitor.
Catalog No. 50-205-3817