Learn More
TARGETMOL CHEMICALS INC CHS-828 25MG
SDP
Catalog No. 502581043
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CHS-828 (GMX1778) a pyridyl cyanoguanidine is an effective inhibitor of NAD+ biosynthesis enzyme NAMPT (IC50 <25 nM). purity: 99%
Catalog No. 50-258-1043 Supplier TARGETMOL CHEMICALS INC Supplier No. T199825MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CHS-828 (GMX1778) a pyridyl cyanoguanidine is an effective inhibitor of NAD+ biosynthesis enzyme NAMPT (IC50 <25 nM). purity: 99%