Learn More
Medchemexpress LLC CHS-828 (GMX1778) | 200484-11-3 | MFCD03700766 | 99.3% | 371.86 g/mol | C19H22ClN5O | 100 MG
SDP
Catalog No. 5000212400
Encompass_Preferred
CHS-828 (GMX1778) is a small-molecule competitive inhibitor of nicotinamide phosphoribosyltransferase (NAMPT) used for research and analytical applications. It lowers cellular NAD+ levels and shows cytotoxic activity in cancer cell models. Supplied as a high-purity analytical standard for laboratory use.
  • Competitive inhibitor of nicotinamide phosphoribosyltransferase (NAMPT).
  • Reduces cellular NAD+ levels leading to cytotoxic effects in cell models.
  • High purity suitable for analytical and research applications.
  • Provided as a solid analytical standard for easy handling and storage.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-12400 Supplier Medchemexpress LLC Supplier No. HY10079100MG
CHS-828 (GMX1778) is a small-molecule competitive inhibitor of nicotinamide phosphoribosyltransferase (NAMPT) used for research and analytical applications. It lowers cellular NAD+ levels and shows cytotoxic activity in cancer cell models. Supplied as a high-purity analytical standard for laboratory use.
  • Competitive inhibitor of nicotinamide phosphoribosyltransferase (NAMPT).
  • Reduces cellular NAD+ levels leading to cytotoxic effects in cell models.
  • High purity suitable for analytical and research applications.
  • Provided as a solid analytical standard for easy handling and storage.
  • Intended for research use only; not for human or clinical use.