Learn More
Aviva Systems Biology Corp Chst11 Antibody - C-terminal region (ARP47444_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47444_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Chst11. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: C4ST1, Chst11ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-306