Learn More
Aviva Systems Biology Corp CHST13 Antibody - C-terminal region (ARP49073_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49073_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHST13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CHST13. Alternate names: C4ST3, MGC119278, MGC119279, MGC119281ApplicationWB. Species reactivity: Guinea Pig, Human
Catalog No. 50-014-60012