Learn More
Aviva Systems Biology Corp CHST14 Antibody - middle region (ARP48994_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48994_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHST14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CHST14. Alternate names: D4ST-1, D4ST1, HD4ST, HNK1ST, ATCSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-014-53569