Learn More
Aviva Systems Biology Corp Chst2 Antibody - middle region (ARP46422_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46422_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Chst2. It was validated on Western Blot. . Alternate names: AI428561, AW121776, C130041E03Rik, Chts2, GST-2, GlcNAc6ST, Gn6stApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-256