Learn More
Aviva Systems Biology Corp CHST4 Antibody - middle region (ARP35787_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35787_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CHST4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human CHST4. Alternate names: GST3, LSST, GlcNAc6ST2, HECGLCNAC6STApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55851