Learn More
Aviva Systems Biology Corp CHST4 Antibody - middle region (ARP35786_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35786_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHST4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CHST4. Alternate names: GST3, LSST, GlcNAc6ST2, HECGLCNAC6STApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Rabbit
Catalog No. 50-014-55850