Learn More
AbD Serotec CHT Ceramic Hydroxyapatite, 100 g
Catalog No. NC1814965
Encompass
100 g, CHT ceramic hydroxyapatite multimodal chromatography media, Type II, 80 um particle size.
Catalog No. NC1814965 Supplier AbD Serotec Supplier No. 1578000
100 g, CHT ceramic hydroxyapatite multimodal chromatography media, Type II, 80 um particle size.