Learn More
Bioworld Chu (N6) Medium, 50 L
Catalog No. 50254563
Encompass_Preferred
Chu (N6) MediumContains Macronutrients and Micronutrients as described by Chu (1975)Provided as 1 unit to make 25L or 50L of medium, or as 10 units that each make 1L of mediumWorking Concentration: 4.0 g/LPlant tissue culture tested Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-563 Supplier Bioworld Supplier No. 306300073
Chu (N6) MediumContains Macronutrients and Micronutrients as described by Chu (1975)Provided as 1 unit to make 25L or 50L of medium, or as 10 units that each make 1L of mediumWorking Concentration: 4.0 g/LPlant tissue culture tested Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested