Learn More
Bioworld Chu (N6) Medium w/ Vitamins, 50 L
Catalog No. 50254566
Encompass_Preferred
Chu (N6) MediumContains Macronutrients, Micronutrients, and Vitamins as described by Chu (1975) Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units that each make 1L of mediumPlant tissue culture testedWorking Concentration: 3.99 g/LpH: 3.5–4.5Soluble in water
Catalog No. 50-254-566 Supplier Bioworld Supplier No. 306300083
Chu (N6) MediumContains Macronutrients, Micronutrients, and Vitamins as described by Chu (1975) Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units that each make 1L of mediumPlant tissue culture testedWorking Concentration: 3.99 g/LpH: 3.5–4.5Soluble in water