Learn More
PHYTOTECHNOLOGY LABS CHU N6 SALTS 100L
Catalog No. NC1789696
Encompass_Preferred
Chu (N6) Medium was developed to promote the initiation, growth, and differentiation of callus from rice pollen cultures.
Catalog No. NC1789696 Supplier PHYTOTECHNOLOGY LABS Supplier No. C416100L
Chu (N6) Medium was developed to promote the initiation, growth, and differentiation of callus from rice pollen cultures.