Learn More
Bioworld Chu Vitamin Concentrate, 10mL Ampules, 100 mL
Catalog No. 50254567
Encompass_Preferred
Chu Vitamin Concentrate (Ampules)Contains the vitamins as described by Chu et al. (1975)Package includes 10 filter sterilized ampules of 10ml of 1000X vitamin stock solution Use 1ml per liter of medium. Contents: Glycine: 2.00mg/LThiamine HCl: 1.0mg/LPyridoxine HCl: 0.5mg/LNicotinic acid: 0.5mg/L
Catalog No. 50-254-567 Supplier Bioworld Supplier No. 306300091
Chu Vitamin Concentrate (Ampules)Contains the vitamins as described by Chu et al. (1975)Package includes 10 filter sterilized ampules of 10ml of 1000X vitamin stock solution Use 1ml per liter of medium. Contents: Glycine: 2.00mg/LThiamine HCl: 1.0mg/LPyridoxine HCl: 0.5mg/LNicotinic acid: 0.5mg/L