Learn More
Bioworld Chu Vitamin Mix, 100 mL
Catalog No. 50254568
Encompass_Preferred
Chu Vitamin MixContains the vitamins as described by Chu et al. (1975).Package includes 0.4 g vitamins to prepare 100ml of 1000X vitamin stock solution Use 1ml per liter of medium.Contents: Glycine: 2.00mg/l Thiamine HCl: 1.0mg/l Pyridoxine HCl: 0.5mg/l Nicotinic acid: 0.5mg/l
Catalog No. 50-254-568 Supplier Bioworld Supplier No. 306300101
Chu Vitamin MixContains the vitamins as described by Chu et al. (1975).Package includes 0.4 g vitamins to prepare 100ml of 1000X vitamin stock solution Use 1ml per liter of medium.Contents: Glycine: 2.00mg/l Thiamine HCl: 1.0mg/l Pyridoxine HCl: 0.5mg/l Nicotinic acid: 0.5mg/l