Learn More
Electron Microscopy Sciences Chuck Adapter for Shandon
SDP
Catalog No. 501900447
Encompass_Preferred
This Chuck Adapter is machined to fit Shandon and Ames / Tissue-Tek cryostats. Chuck Adapter machined to fit Shandon cryostats.
Catalog No. 50-190-0447 Supplier Electron Microscopy Sciences Supplier No. 6173101
This Chuck Adapter is machined to fit Shandon and Ames / Tissue-Tek cryostats. Chuck Adapter machined to fit Shandon cryostats.