Learn More
PATHOLOGY INNOVATIONS CHUCK BIN
Catalog No. NC1661234
Encompass_Preferred
A sturdy little stainless steel box to shelter chucks while staying cold in the cryostat. Loose fitting unattached cover for easy access.
Catalog No. NC1661234 Supplier PATHOLOGY INNOVATIONS Supplier No. 142
A sturdy little stainless steel box to shelter chucks while staying cold in the cryostat. Loose fitting unattached cover for easy access.