Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caframo™ Chuck Guard Accessory with Magnetic Enclosure

Catalog No. 14501101 Shop All Caframo Products
Change view
Click to view available options
Type:
Chuck Guard
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14-501-101 Chuck Guard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-501-101 Supplier Caframo™ Supplier No. CG1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chuck Guard safely encloses the spinning chuck. Magnets snap the guard in place. Can be used with the following stirrers: BDC1850 (14-500-10), BDC3030 (14-500-11), BDC6015 (14-500-12) and FSR3000 (14-500-210).

  • Easy to install and remove - magnets snap in place
  • Creates a barrier around the spinning chuck
  • Protects the chuck from splashes

Specifications

Color Clear
For Use With (Equipment) Caframo™ and Fisherbrand™ stirrers
Includes Chuck guard with two installed magnets
Type Chuck Guard
Weight (Metric) 14 g
Material Polyurethane
Weight (English) 0.5 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.