Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caframo Limited™ Chuck Guard Kit with Adapter

Catalog No. 14500573 Shop All Caframo Products
Change view
Click to view available options
Color:
Clear, Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-500-573 Clear, Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-500-573 Supplier Caframo Limited™ Supplier No. CG1KIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes CG1 chuck guard+ adapter

  • Implement safety with a quick snap of this clear chemical-resistant polycarbonate chuck guard to your Caframo Light Torque stirrer (BDC2002 Compact, BDC2010 Reversing Digital, and Fisherbrand™ compact digital overhead stirrer) housing using the included adapter
  • Creates a barrier around the moving chuck, as well as helps protect the chuck area from chemical splashes
  • Easy to attach and remove

Specifications

Color Clear, Gray
For Use With (Equipment) BDC2002, BDC2010, Fisherbrand™ Compact Digital Overhead Stirrer
For Use With (Application) For use with Caframo™ and Fisherbrand™ Light torque overhead stirrers
Type Chuck Guard Kit
Material Polycarbonate, Vinyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.