Learn More
Arrow Engineering Adjustable Keyless Chuck
Catalog No. NC1721852
Encompass_Preferred
With a 3/8"dia. Motor shaft adapter the Arrow Mixing Products keyless chucks are a popular choice thanks to their precision, ease of use, and dependable performance—perfect for a variety of lab applications. Our KLC50-375 has a capacity from 1/16″ to 1/2″ dia. (1.5 mm to 12.7 mm), offering the flexibility you need for precise and efficient mixing.
Catalog No. NC1721852 Supplier Arrow Engineering Supplier No. KLC50375
With a 3/8"dia. Motor shaft adapter the Arrow Mixing Products keyless chucks are a popular choice thanks to their precision, ease of use, and dependable performance—perfect for a variety of lab applications. Our KLC50-375 has a capacity from 1/16″ to 1/2″ dia. (1.5 mm to 12.7 mm), offering the flexibility you need for precise and efficient mixing.