Learn More
Electron Microscopy Sciences PolarChuck™ A, Avantik/Microm, Waffle Grid, 40 mm
SDP
Catalog No. 501928642
Encompass_Preferred
PolarChuck cryostat embedding chucks are made of anodized aluminum. Deep rectangular waffle grid or circular pattern creates a more stable and secure tissue mount.Size: 40 mm
Catalog No. 50-192-8642 Supplier Electron Microscopy Sciences Supplier No. 7017106
PolarChuck cryostat embedding chucks are made of anodized aluminum. Deep rectangular waffle grid or circular pattern creates a more stable and secure tissue mount.Size: 40 mm