Learn More
MSC Chums Eyewear Retainer, Cotton, Black, Single Breakaway, 1 ea
Catalog No. 5000768596
Encompass
This cotton eyewear retainer is designed for safety in environments with choking or snagging hazards. It features a single breakaway mechanism that separates with 6 lb. of pressure and can be easily reattached for continued use.
  • Breakaway for safety behind the neck
  • Reattaches for continued use
  • Compatible with most standard eyewear frames
  • Made in the USA
Catalog No. 50-007-68596 Supplier MSC Supplier No. 67035345
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This cotton eyewear retainer is designed for safety in environments with choking or snagging hazards. It features a single breakaway mechanism that separates with 6 lb. of pressure and can be easily reattached for continued use.
  • Breakaway for safety behind the neck
  • Reattaches for continued use
  • Compatible with most standard eyewear frames
  • Made in the USA