Learn More
MSC Chums Eyewear Retainer, Flame Resistant Single Breakaway, Kevlar, 1 ea
Catalog No. 5000673833
Encompass
This flame-resistant eyewear retainer is engineered for demanding work environments, providing reliable eyewear retention without compromising safety. Constructed from Kevlar, it features a breakaway design and is resistant to ignition, melting, and dripping when exposed to flames or arc flashes.
  • Breakaway design for safety and reattachment
  • Flame resistant and arc-rated to 15 cal/square cm
  • Resistant to direct flame, sparks, and welding spatter
  • Kevlar integrity maintained up to 450℃
  • Compatible with all major safety eyewear brands
  • Meets NFPA 70E standards
  • Usable length adjustable to 24 in
Catalog No. 50-006-73833 Supplier MSC Supplier No. 54547070
This flame-resistant eyewear retainer is engineered for demanding work environments, providing reliable eyewear retention without compromising safety. Constructed from Kevlar, it features a breakaway design and is resistant to ignition, melting, and dripping when exposed to flames or arc flashes.
  • Breakaway design for safety and reattachment
  • Flame resistant and arc-rated to 15 cal/square cm
  • Resistant to direct flame, sparks, and welding spatter
  • Kevlar integrity maintained up to 450℃
  • Compatible with all major safety eyewear brands
  • Meets NFPA 70E standards
  • Usable length adjustable to 24 in