Learn More
MSC Chums Mono Orbiter, Monofilament, Lime Yellow, 15.5 in, 1 ea
Catalog No. 5000768095
Encompass
The Mono Orbiter is a strong, sleek, and lightweight eyewear retainer designed to securely keep glasses on the wearer's head. Its high-tenacity monofilament construction offers stiffness, preventing draping around the neckline or interference with headwear.
  • Low-profile Orbiter ends
  • Precision-molded rubber ends accommodate various frames for reliable grip
  • Made from lightweight, high-tenacity monofilament
  • Total length 15.5 in
  • Non-heat conductive
  • Compatible with most safety glasses
  • Meets NFPA 70E standards
  • Made in the USA
Catalog No. 50-007-68095 Supplier MSC Supplier No. 79571055
The Mono Orbiter is a strong, sleek, and lightweight eyewear retainer designed to securely keep glasses on the wearer's head. Its high-tenacity monofilament construction offers stiffness, preventing draping around the neckline or interference with headwear.
  • Low-profile Orbiter ends
  • Precision-molded rubber ends accommodate various frames for reliable grip
  • Made from lightweight, high-tenacity monofilament
  • Total length 15.5 in
  • Non-heat conductive
  • Compatible with most safety glasses
  • Meets NFPA 70E standards
  • Made in the USA