Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chums™ Original Cotton Standard End Eyewear Retainers

Catalog No. 19808671 Shop All Chums Ltd Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Color:
Assorted in Pack of 25
Purple
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-808-671 Assorted in Pack of 25
19-058-607 Purple
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-808-671 Supplier Chums™ Supplier No. 1211514
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

World's first 100% cotton, fully adjustable eyewear retainer

World's first 100% cotton, fully adjustable eyewear retainer

Tough and durable, yet soft enough for everyday use.

  • Fits most standard frames
  • Logo option available
  • Made in the USA

Order Info

Available in a variety of popular colors.

Specifications

Color Assorted in Pack of 25
Product Type Eyewear Retainer
For Use With (Equipment) Fits most standard frames
No. per Pack 200/Pk.
Material Cotton
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.