Learn More
Innovative Research Inc Chx RBCs Packed 5 100ml
Catalog No. 502744614
Encompass_Preferred
Chicken Red Blood Cells Packed 5% 100ml - Chicken Red Blood Cells from Innovative Research are manufactured using whole blood collections from chickens. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 5% solution in Alsever's. Red blood cells are perishable and are collected and processed upon receipt of order.
Catalog No. 50-274-4614 Supplier Innovative Research Inc Supplier No. ICHRBC5P100ML
Chicken Red Blood Cells Packed 5% 100ml - Chicken Red Blood Cells from Innovative Research are manufactured using whole blood collections from chickens. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 5% solution in Alsever's. Red blood cells are perishable and are collected and processed upon receipt of order.