Learn More
Medchemexpress LLC HY-112951 10mg Medchemexpress, ChX710 CAS: Purity:>98%
SDP
Catalog No. 501969212
Encompass_Preferred
Medchemexpress, HY-112951 10mg ChX710 CAS: ChX710 could prime the type I interferon response to cytosolic DNA, which induces the ISRE promoter sequence, specific cellular Interferon-Stimulated Genes (ISGs), and the phosphorylation of Interferon Regulatory Factor (IRF) 3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9212 Supplier Medchemexpress LLC Supplier No. HY11295110MG
Medchemexpress, HY-112951 10mg ChX710 CAS: ChX710 could prime the type I interferon response to cytosolic DNA, which induces the ISRE promoter sequence, specific cellular Interferon-Stimulated Genes (ISGs), and the phosphorylation of Interferon Regulatory Factor (IRF) 3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.