Learn More
Athens Research And Technology Inc CHYLOMICRONS HU 1MG
Supplier:  Athens Research And Technology Inc 12160308251MG
Encompass_Preferred
Chylomicrons, Human Plasma: 1 MG; Liquid in 150 mM NaCl, pH 7.4, and 0.01% EDTA; Density: <0.95 g/ml; Composition: 98% lipid, 2% protein. This native protein purified from fresh, non-frozen plasma in the Athens Research & Tech lab. Lipoprotein
Catalog No. 50-489-711