Learn More
Alkali Scientific Chymostatin, 1 Milligram
SDP
Catalog No. 5000660235
Encompass_Preferred
Chymostatin is a potent protease inhibitor used in laboratory research to inhibit serine and cysteine proteases, including chymotrypsin and other enzymes involved in protein degradation. This 1-milligram vial is ideal for small-scale experiments requiring the prevention of proteolytic activity. Chymostatin is widely used in enzyme inhibition assays, protein research, and the analysis of enzyme-substrate interactions. It is particularly useful in proteomics to ensure that proteins of interest remain intact during extraction and analysis. This reagent is commonly applied in studies on enzyme function, protein stability, and enzymatic reactions in biological systems.
Catalog No. 50-006-60235 Supplier Alkali Scientific Supplier No. CHY002CP
Chymostatin is a potent protease inhibitor used in laboratory research to inhibit serine and cysteine proteases, including chymotrypsin and other enzymes involved in protein degradation. This 1-milligram vial is ideal for small-scale experiments requiring the prevention of proteolytic activity. Chymostatin is widely used in enzyme inhibition assays, protein research, and the analysis of enzyme-substrate interactions. It is particularly useful in proteomics to ensure that proteins of interest remain intact during extraction and analysis. This reagent is commonly applied in studies on enzyme function, protein stability, and enzymatic reactions in biological systems.