Learn More
Carbosynth LLC. Chymostatin, 100mg, Mixture of Type A, B, and C[(S)-1-Carboxy-2-phenylethyl]-carbamoyl-a-[2-iminohexahydro-4(S)-pyrimidyl]-(S)-glycyl-X-phenylalaninal X: L-leucyl (Type A), L-valyl (Type B), L-isoleucyl (Type C), MW: 610.72, Molecular formula: C31H42N6O7, CAS# 9076-44-2
SDP
Catalog No. 501685844
Encompass_Preferred
Synonyms: [(S)-1-Carboxy-2-phenylethyl]carbamoyl-a-[2-iminohexahydro-4(S)-pyrimidyl]-(S)-glycyl-X-phenylalaninal, Application: Microbial Product Inhibitor for Chymotrypsin, Papain, and Cathepsin B/G, Storage Temperature: -20°C, References: H. Umezawa, T. Aoyagi, H. Morishima, S. Kunimoto, M. Matsuzaki, M. Hamada, and T. Takeuchi, J. Antibiotics, 23, 425 (1970). (Original), K. Tatsuta, N. Mikami, K. Fujimoto, S. Umezawa, H. Umezawa, and T. Aoyagi, J. Antibiotics, 26, 625 (1973). (Chem. Structure), L.A. Johnson, K.E. Moon, and M. Eisenberg, Biochim. Biophys. Acta, 953, 269 (1988). (Inhibitory Activity)
Catalog No. 50-168-5844 Supplier Carbosynth LLC. Supplier No. ICY4063100MG
Synonyms: [(S)-1-Carboxy-2-phenylethyl]carbamoyl-a-[2-iminohexahydro-4(S)-pyrimidyl]-(S)-glycyl-X-phenylalaninal, Application: Microbial Product Inhibitor for Chymotrypsin, Papain, and Cathepsin B/G, Storage Temperature: -20°C, References: H. Umezawa, T. Aoyagi, H. Morishima, S. Kunimoto, M. Matsuzaki, M. Hamada, and T. Takeuchi, J. Antibiotics, 23, 425 (1970). (Original), K. Tatsuta, N. Mikami, K. Fujimoto, S. Umezawa, H. Umezawa, and T. Aoyagi, J. Antibiotics, 26, 625 (1973). (Chem. Structure), L.A. Johnson, K.E. Moon, and M. Eisenberg, Biochim. Biophys. Acta, 953, 269 (1988). (Inhibitory Activity)