Learn More
Cayman Chemical ChymostatIn 25mg
SDP
Catalog No. 502973019
Encompass_Preferred
A potent inhibitor of chymotrypsin and chymase (Ki = 9.36 and 13.1 nM, respectively), that less effectively blocks the activity of cathepsins, papain, and leukocyte elastase
Catalog No. 50-297-3019 Supplier Cayman Chemical Supplier No. 1511425 MG
A potent inhibitor of chymotrypsin and chymase (Ki = 9.36 and 13.1 nM, respectively), that less effectively blocks the activity of cathepsins, papain, and leukocyte elastase