Learn More
Research Products International Corp Chymostatin, 5 Milligrams
SDP
Catalog No. 50488590
Encompass_Preferred
Reversible serine protease inhibitor of a,b,y, and chymostatin, papain, and cathepsins A,B,C,.
Working concentrations: 10-100uM.
Catalog No. 50-488-590 Supplier Research Products International Corp Supplier No. C400250.005
Reversible serine protease inhibitor of a,b,y, and chymostatin, papain, and cathepsins A,B,C,.
Working concentrations: 10-100uM.