Learn More
Cayman Chemical Chymostatin, Purity: ≥95% (a mixture of A, B, C), CAS: 9076-44-2, 50 mg
SDP
Catalog No. 502969699
Encompass_Preferred
A potent inhibitor of chymotrypsin and chymase (Ki = 9.36 and 13.1 nM, respectively), that less effectively blocks the activity of cathepsins, papain, and leukocyte elastase
Catalog No. 50-296-9699 Supplier Cayman Chemical Supplier No. 1511450 MG
A potent inhibitor of chymotrypsin and chymase (Ki = 9.36 and 13.1 nM, respectively), that less effectively blocks the activity of cathepsins, papain, and leukocyte elastase