Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chymostatin MP Biomedicals

Catalog No. ICN15284501 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15284501 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15284501 Supplier MP Biomedicals, Inc Supplier No. 0215284501
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9076-44-2
Molecular Formula C31H41N7O6
Molecular Weight (g/mol) 607.712
InChI Key MRXDGVXSWIXTQL-MWVOZXJSSA-N
Synonym chymostatin
PubChem CID 129894295
IUPAC Name (2S)-2-[[(1S)-1-(2-amino-1,4,5,6-tetrahydropyrimidin-6-yl)-2-[[4-methyl-1-oxo-1-[[(2S)-1-oxo-3-phenylpropan-2-yl]amino]pentan-2-yl]amino]-2-oxoethyl]carbamoylamino]-3-phenylpropanoic acid
SMILES CC(C)CC(C(=O)NC(CC1=CC=CC=C1)C=O)NC(=O)C(C2CCN=C(N2)N)NC(=O)NC(CC3=CC=CC=C3)C(=O)O

Specifications

Quantity 1 mg
Chemical Name or Material Chymostatin
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.