Learn More
Athens Research And Technology Inc CHYMOTRYPSIN HU PANCREAS 100UG
Supplier:  Athens Research And Technology Inc 1619030820100UG
Encompass_Preferred
Chymotrypsin, Human Pancreas: 100 UG; Salt-free lyophilized solid. MW 25,000; EC=1.9; Activity: 40-70 units per mg protein; Purity: > =95% by SDS-PAGE. This native protein is prepared and purified from human tissue in the Athens Research & Tech lab.
Catalog No. 50-489-714