Learn More
CPC Scientific Suc-Leu-Leu-Val-Tyr-AMC 1MG
SDP
Supplier:  CPC Scientific SUBS012A
Encompass
Fluorogenic peptide used in the analysis of chymotrypsin-like activity of trypsins in addition to chymase from rat mast cells. Furthermore, it is a substrate for a chymotrpsin-like enzyme from sperm extract of the ascidian Halocynthia roretzi and a strong inhibitor of fertilization. It is also a substrate for carboxypeptidase Y, proteinase yscE, porcine calpain isozymes I and II, papain, and the multicatalytic proteinase, ingensin.SEQUENCE: Suc-Leu-Leu-Val-Tyr-AMCONE-LETTER SEQUENCE: Suc-LLVY-AMCMOLECULAR FORMULA: C40H53N5O10MOLECULAR WEIGHT: 763.89STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [94367-21-2]SYNONYMS: Suc-Leu-Leu-Val-Tyr-AMCRESEARCH AREA: ApoptosisREFERENCES:SKU(s): SUBS-012A, SUBS-012B, SUBS-012C, SUBS-012D
Catalog No. 50-211-0482