Learn More
Worthington Biochemical Corporation Chymotrypsin, Alpha, TLCK Treated, Sequence, 4x100 ug
SDP
Supplier:  Worthington Biochemical Corporation LS02132
Encompass
Three time crystallized and treated with 1-chloro-3-tosylamido-7-amino-2-heptanone (TLCK) to inhibit trypsin activity [Shaw, et al., Biochemistry, 4, 2219 (1965)]. Dialyzed to remove autolysis products and low molecular weight contaminants. Supplied as lyophilized powder. Store at 2-8°C.
Catalog No. NC1624395