Learn More
Worthington Biochemical Corporation Chymotrypsinogen A, 5 g
SDP
Supplier:  Worthington Biochemical Corporation LS005623
Encompass
5X crystallized, electrophoretically homogeneous. Supplied as a dialyzed, lyophilized powder.
Catalog No. NC1553704