Learn More
Thermo Scientific Chemicals Chymotrypsinogen A, from bovine pancreas
Description
Chymotrypsinogen A is used as a serine protease that hydrolyzes peptide bonds with aromatic or large hydrophobic side chains (Tyr, Trp, Phe, Met) on the carboxyl end of the peptide bond, used in the non-invasive determination of solid-state protein conformation using near infrared (NIR) spectroscopy. It has been used to study the partitioning of protein in polymer/polymer aqueous two-phase systems. The enzyme has also been used for self-interaction chromatography applications, to test the rapid measurement of protein osmotic second virial coefficients. In this technique, the protein is immobilized on chromatographic particles and its retention is measured using isocratic elution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| For Use With (Application) | The zymogen of chymotrypsin |
| Molecular Weight (g/mol) | 25.6 kd |
| Content And Storage | -30°C to -10°C |
| Form | Powder |
| Quantity | 1 g |
| Product Type | Protease |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.