Learn More
TARGETMOL CHEMICALS INC CI-1040 25MG
SDP
Catalog No. 502580809
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CI-1040 (PD 184352) (PD184352) is an ATP non-competitive MEK1/2 inhibitor (IC50 17 nM). purity: 99%
Catalog No. 50-258-0809 Supplier TARGETMOL CHEMICALS INC Supplier No. T244325MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CI-1040 (PD 184352) (PD184352) is an ATP non-competitive MEK1/2 inhibitor (IC50 17 nM). purity: 99%