Learn More
Selleck Chemical LLC PD184352 (CI-1040) 25mg 212631-79-3
SDP
Catalog No. 508716
Encompass_Preferred
PD184352 (CI-1040) is an ATP non-competitive MEK1/2 inhibitor with IC50 of 17 nM in cell-based assays, 100-fold more selective for MEK1/2 than MEK5. PD184352 (CI-1040) selectively induces apoptosis. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-871-6 Supplier Selleck Chemical LLC Supplier No. S102025MG
PD184352 (CI-1040) is an ATP non-competitive MEK1/2 inhibitor with IC50 of 17 nM in cell-based assays, 100-fold more selective for MEK1/2 than MEK5. PD184352 (CI-1040) selectively induces apoptosis. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.