Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ciba Blue 2B, TCI America™
SDP

Catalog No. T056225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T056225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T056225G Supplier TCI America Supplier No. T056225G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2475-31-2
Molecular Formula C16H6Br4N2O2
Molecular Weight (g/mol) 577.85
MDL Number MFCD00049382
InChI Key PTWYQANXSNMUTI-YPKPFQOOSA-N
Synonym 5,5′C,7,7′C-Tetrabromoindigo, Vat Blue 5
PubChem CID 5285390
IUPAC Name (Z)-5,5',7,7'-tetrabromo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-3,3'-dione
SMILES BrC1=CC(Br)=C2N\C(C(=O)C2=C1)=C1/NC2=C(Br)C=C(Br)C=C2C1=O

Specifications

Color Blue-Purple
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Ciba Blue 2B
EINECSNumber (5)-2228
RTECSNumber NM3259100
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.